accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNHSNHNH2

CAS RN 32813-48-2

IUPAC Name: 3-amino-1-[3-(morpholin-4-yl)propyl]thiourea
Synonyms: 3-amino-1-[3-(morpholin-4-yl)propyl]thiourea
Molecular Weight (g/mol): 218.32
Molecular Formula: C8H18N4OS
InChi Key: CDEOWFKHBKXPIO-UHFFFAOYSA-N
SMILES: NNC(=S)NCCCN1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results