Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-[3-(4-Morpholinyl)propyl]-3-thiosemicarbazide, 98+%, Thermo Scientific™

Catalog No. AAL1071403
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1071403 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1071403 Supplier Thermo Scientific Chemicals Supplier No. L1071403

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 32813-48-2
Molecular Formula C8H18N4OS
Quantity 1g
MDL Number MFCD00023400
Beilstein 881684
Synonym 4-3-morpholinopropyl-3-thiosemicarbazide, n-3-morpholinopropyl hydrazinecarbothioamide, 4-3-4-morpholino propyl-3-thiosemicarbazide, 3-amino-1-3-morpholin-4-yl propyl thiourea, 4-3-4-morpholinyl propyl-3-thiosemicarbazide, n-3-morpholin-4-yl propyl hydrazinecarbothioamide, maybridge1_006942, hydrazino 3-morpholin-4-ylpropyl amino methane-1-thione, 1-amino-3-3-morpholin-4-ylpropyl thiourea, 1-amino-3-3-4-morpholinyl propyl thiourea
InChI Key CDEOWFKHBKXPIO-UHFFFAOYSA-N
SMILES C1COCCN1CCCNC(=S)NN
IUPAC Name 1-amino-3-(3-morpholin-4-ylpropyl)thiourea
Molecular Weight (g/mol) 218.319
PubChem CID 1809200
Formula Weight 218.32
Percent Purity ≥98%
Chemical Name or Material 4-[3-(4-Morpholinyl)propyl]-3-thiosemicarbazide
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.