accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3282-30-2

IUPAC Name: 2,2-dimethylpropanoyl chloride
Synonyms: 2,2-dimethylpropanoyl chloride
Molecular Weight (g/mol): 120.58
Molecular Formula: C5H9ClO
InChi Key: JVSFQJZRHXAUGT-UHFFFAOYSA-N
SMILES: CC(C)(C)C(Cl)=O
Boiling Point 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)