accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32829-08-6

IUPAC Name: 3,6,7,10,11-pentakis(acetyloxy)triphenylen-2-yl acetate
Synonyms: 3,6,7,10,11-pentakis(acetyloxy)triphenylen-2-yl acetate
Molecular Weight (g/mol): 576.51
Molecular Formula: C30H24O12
InChi Key: YTKHQSUTXVUOGK-UHFFFAOYSA-N
SMILES: CC(=O)OC1=C(OC(C)=O)C=C2C(=C1)C1=CC(OC(C)=O)=C(OC(C)=O)C=C1C1=CC(OC(C)=O)=C(OC(C)=O)C=C21

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results