Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,6,7,10,11-Hexaacetoxytriphenylene, TCI America™
SDP

Catalog No. H0923100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0923100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H0923100MG Supplier TCI America Supplier No. H0923100MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 32829-08-6
Molecular Formula C30H24O12
Molecular Weight (g/mol) 576.51
MDL Number MFCD01321173
InChI Key YTKHQSUTXVUOGK-UHFFFAOYSA-N
PubChem CID 11827967
IUPAC Name (3,6,7,10,11-pentaacetyloxytriphenylen-2-yl) acetate
SMILES CC(=O)OC1=C(C=C2C(=C1)C3=CC(=C(C=C3C4=CC(=C(C=C24)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Quantity 100 mg
Formula Weight 576.51
Chemical Name or Material 2,3,6,7,10,11-Hexaacetoxytriphenylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.