accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32854-09-4

IUPAC Name: dihydrogen methyl 2-amino-3-[(2-amino-3-methoxy-3-oxopropyl)disulfanyl]propanoate dichloride
Synonyms: dihydrogen methyl 2-amino-3-[(2-amino-3-methoxy-3-oxopropyl)disulfanyl]propanoate dichloride
Molecular Weight (g/mol): 341.26
Molecular Formula: C8H18Cl2N2O4S2
InChi Key: QKWGUPFPCRKKMQ-UHFFFAOYNA-N
SMILES: [H+].[H+].[Cl-].[Cl-].COC(=O)C(N)CSSCC(N)C(=O)OC
Quantity 
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)