Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Apn-Maleimide

Catalog No. 80653650MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-653-650MG 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 80-653-650MG Supplier Sigma Aldrich Supplier No. 80653650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

APN-Maleimide is a bifunctional crosslinker for thiol-to-thiol coupling. The coupling can be performed with high selectivity in biological medium using mild reaction conditions. Due to kinetic resolution the first thiol reacts exclusively with the maleimide residue producing the protein-APN conjugate. This conjugate can be readily coupled with thiol-containing molecules.

Specifications

CAS 32854-09-4
Melting Point 182°C to 183°C
Linear Formula CH3OCOCH(NH2)CH2SSCH2CH(NH2)COOCH3 -+ 2HCl
MDL Number MFCD00012490
Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.