CAS RN 32863-32-4
CAS RN 32863-32-4
IUPAC Name:
2,1,3-benzoxadiazole-4-carbaldehyde
Synonyms:
2,1,3-benzoxadiazole-4-carbaldehyde
Molecular Weight (g/mol):
148.12
Molecular Formula:
C7H4N2O2
InChi Key:
YBBRQAXNTWMMFZ-UHFFFAOYSA-N
SMILES:
O=CC1=CC=CC2=NON=C12
1
–
1
of
1
results
1
2,1,3-Benzoxadiazole-4-carboxaldehyde 98.0+%, TCI America™
CAS: 32863-32-4 Molecular Formula: C7H4N2O2 Molecular Weight (g/mol): 148.12 MDL Number: MFCD07772900 InChI Key: YBBRQAXNTWMMFZ-UHFFFAOYSA-N Synonym: 4-Benzofurazancarboxaldehyde, 4-Formylbenzofurazan PubChem CID: 10290873 IUPAC Name: 2,1,3-benzoxadiazole-4-carbaldehyde SMILES: O=CC1=CC=CC2=NON=C12