Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,1,3-Benzoxadiazole-4-carboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. B49281G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B49281G Supplier TCI America Supplier No. B49281G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 32863-32-4
Molecular Formula C7H4N2O2
Molecular Weight (g/mol) 148.12
MDL Number MFCD07772900
InChI Key YBBRQAXNTWMMFZ-UHFFFAOYSA-N
Synonym 4-Benzofurazancarboxaldehyde, 4-Formylbenzofurazan
PubChem CID 10290873
IUPAC Name 2,1,3-benzoxadiazole-4-carbaldehyde
SMILES O=CC1=CC=CC2=NON=C12

Specifications

Melting Point 108°C
Color Yellow
Quantity 1 g
Formula Weight 148.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,1,3-Benzoxadiazole-4-carboxaldehyde
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.