accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3289-50-7

IUPAC Name: 2,6-dimethoxypyrimidin-4-amine
Synonyms: 2,6-dimethoxypyrimidin-4-amine
Molecular Weight (g/mol): 155.16
Molecular Formula: C6H9N3O2
InChi Key: LNTJJKHTAZFVJJ-UHFFFAOYSA-N
SMILES: COC1=CC(N)=NC(OC)=N1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)