Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2,6-dimethoxypyrimidine, 98%, Thermo Scientific™

Catalog No. AAH6619014
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6619014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6619014 Supplier Thermo Scientific Chemicals Supplier No. H6619014

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3289-50-7
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.157
MDL Number MFCD00082802
InChI Key LNTJJKHTAZFVJJ-UHFFFAOYSA-N
Synonym 4-amino-2,6-dimethoxypyrimidine, 4-pyrimidinamine, 2,6-dimethoxy, 6-amino-2,4-dimethoxypyrimidine, 2,6-dimethoxy-4-aminopyrimidine, 2,6-dimethoxy-pyrimidin-4-ylamine, 2,4-dimethoxy-6-aminopyrimidine, 2,6-dimethoxy-4-pyrimidinamine, pyrimidine, 4-amino-2,6-dimethoxy, 2,6-dimethoxypyrimidine-4-ylamine, 2,6-dimethoxypyrimidin-4-amin
PubChem CID 76789
IUPAC Name 2,6-dimethoxypyrimidin-4-amine
SMILES COC1=NC(=NC(=C1)N)OC

Specifications

Melting Point 149°C to 152°C
Color White
Quantity 25 g
Beilstein 137580
Formula Weight 155.16
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 4-Amino-2,6-dimethoxypyrimidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-946-2
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.