CAS RN 3290-01-5
IUPAC Name:
1,2-dichloro-3-(chloromethyl)benzene
Synonyms:
1,2-dichloro-3-(chloromethyl)benzene
Molecular Weight (g/mol):
195.47
Molecular Formula:
C7H5Cl3
InChi Key:
AIPJZPPOFWCJRC-UHFFFAOYSA-N
SMILES:
ClCC1=CC=CC(Cl)=C1Cl
1
–
1
of
1
results
1
2,3-Dichlorobenzyl Chloride 98.0+%, TCI America™
CAS: 3290-01-5 Molecular Formula: C7H5Cl3 Molecular Weight (g/mol): 195.47 MDL Number: MFCD00035817 InChI Key: AIPJZPPOFWCJRC-UHFFFAOYSA-N Synonym: alpha,2,3-Trichlorotoluene PubChem CID: 76790 IUPAC Name: 1,2-dichloro-3-(chloromethyl)benzene SMILES: C1=CC(=C(C(=C1)Cl)Cl)CCl