Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichlorobenzyl Chloride 98.0+%, TCI America™
SDP

Catalog No. D20605G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20605G 5 g
D206025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20605G Supplier TCI America Supplier No. D20605G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3290-01-5
Molecular Formula C7H5Cl3
Molecular Weight (g/mol) 195.47
MDL Number MFCD00035817
InChI Key AIPJZPPOFWCJRC-UHFFFAOYSA-N
Synonym alpha,2,3-Trichlorotoluene
PubChem CID 76790
IUPAC Name 1,2-dichloro-3-(chloromethyl)benzene
SMILES C1=CC(=C(C(=C1)Cl)Cl)CCl

Specifications

Color Yellow
Boiling Point 255°C
Quantity 5 g
UN Number 3265
Formula Weight 195.47
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Dichlorobenzyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.