accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH2

CAS RN 3295-97-4

IUPAC Name: 1-(prop-2-en-1-yloxy)octane
Synonyms: 1-(prop-2-en-1-yloxy)octane
Molecular Weight (g/mol): 170.3
Molecular Formula: C11H22O
InChi Key: IELYMBBIHQDONA-UHFFFAOYSA-N
SMILES: CCCCCCCCOCC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results