accessibility menu, dialog, popup

UserName

Viewing profile as user:
N

CAS RN 3297-72-1

IUPAC Name: 1-phenylisoquinoline
Synonyms: 1-phenylisoquinoline
Molecular Weight (g/mol): 205.26
Molecular Formula: C15H11N
InChi Key: LPCWDYWZIWDTCV-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=C2C=CC=CC2=CC=N1
Boiling Point 
  • (2)
  • (1)
Physical Form 
  • (3)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results