accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHONOOCl

CAS RN 329940-70-7

IUPAC Name: 2-chloro-N-(3-methylphenyl)-4-nitrobenzamide
Synonyms: 2-chloro-N-(3-methylphenyl)-4-nitrobenzamide
Molecular Weight (g/mol): 290.7
Molecular Formula: C14H11ClN2O3
InChi Key: VIQSGUZRECTJEM-UHFFFAOYSA-N
SMILES: CC1=CC(NC(=O)C2=C(Cl)C=C(C=C2)[N+]([O-])=O)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results