Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-N-(3-methylphenyl)-4-nitrobenzamide, 97%, Thermo Scientific™

Catalog No. AAH59987MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59987MD 250 mg
AAH5998703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH59987MD Supplier Thermo Scientific Chemicals Supplier No. H59987MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 329940-70-7
Molecular Formula C14H11ClN2O3
Molecular Weight (g/mol) 290.703
MDL Number MFCD00784494
InChI Key VIQSGUZRECTJEM-UHFFFAOYSA-N
Synonym 2-chloro-n-3-methylphenyl-4-nitrobenzamide, 2-chloro-4-nitro-n-3-methylphenyl benzamide, cambridge id 5344357, 2-chloro-4-nitrophenyl-n-3-methylphenyl carboxamide
PubChem CID 710885
IUPAC Name 2-chloro-N-(3-methylphenyl)-4-nitrobenzamide
SMILES CC1=CC(=CC=C1)NC(=O)C2=C(C=C(C=C2)[N+](=O)[O-])Cl

Specifications

Quantity 250 mg
Formula Weight 290.7
Percent Purity 97%
Chemical Name or Material 2-Chloro-N-(3-methylphenyl)-4-nitrobenzamide
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.