accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 330-14-3

IUPAC Name: 4-[(trifluoromethyl)sulfanyl]benzoyl chloride
Synonyms: 4-[(trifluoromethyl)sulfanyl]benzoyl chloride
Molecular Weight (g/mol): 240.62
Molecular Formula: C8H4ClF3OS
InChi Key: BCMFTOCCLOAGBP-UHFFFAOYSA-N
SMILES: FC(F)(F)SC1=CC=C(C=C1)C(Cl)=O
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)