accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNF

CAS RN 330-83-6

IUPAC Name: 4-fluoro-N-phenylaniline
Synonyms: 4-fluoro-N-phenylaniline
Molecular Weight (g/mol): 187.22
Molecular Formula: C12H10FN
InChi Key: RBWCFTJQRDTIBX-UHFFFAOYSA-N
SMILES: FC1=CC=C(NC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results