Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorodiphenylamine 98.0+%, TCI America™
SDP

Catalog No. F06305G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06305G 5 g
F06301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06305G Supplier TCI America Supplier No. F06305G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 330-83-6
Molecular Formula C12H10FN
Molecular Weight (g/mol) 187.217
MDL Number MFCD01318538
InChI Key RBWCFTJQRDTIBX-UHFFFAOYSA-N
Synonym N-(4-Fluorophenyl)aniline
PubChem CID 2774468
IUPAC Name 4-fluoro-N-phenylaniline
SMILES C1=CC=C(C=C1)NC2=CC=C(C=C2)F

Specifications

Melting Point 38°C
Boiling Point 166°C
Quantity 5 g
Formula Weight 187.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorodiphenylamine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.