accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONHNOOH3C

CAS RN 3304-70-9

IUPAC Name: 4,5-dimethyl 1H-imidazole-4,5-dicarboxylate
Synonyms: 4,5-dimethyl 1H-imidazole-4,5-dicarboxylate
Molecular Weight (g/mol): 184.15
Molecular Formula: C7H8N2O4
InChi Key: CUIWFAXEALIQJS-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N=CN1)C(=O)OC
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)