Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl 1H-Imidazole-4,5-dicarboxylate 98.0+%, TCI America™
SDP

Catalog No. I045625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I045625G 25 g
I045610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I045625G Supplier TCI America Supplier No. I045625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3304-70-9
Molecular Formula C7H8N2O4
Molecular Weight (g/mol) 184.151
MDL Number MFCD00051338
InChI Key CUIWFAXEALIQJS-UHFFFAOYSA-N
Synonym dimethyl 4,5-imidazoledicarboxylate, 1h-imidazole-4,5-dicarboxylic acid dimethyl ester, dimethyl imidazole-4,5-dicarboxylate, 1h-imidazole-4,5-dicarboxylic acid, dimethyl ester, 4,5-dimethyl 1h-imidazole-4,5-dicarboxylate, imidazole-4,5-dicarboxylic acid, dimethyl ester, dimethyl 4,5-imidazole-dicarboxylate, 1h-imidazole-4,5-dicarboxylic acid, 4,5-dimethyl ester, imidazole-4, dimethyl ester, 1h-imidazole-4, dimethyl ester
PubChem CID 76810
IUPAC Name dimethyl 1H-imidazole-4,5-dicarboxylate
SMILES COC(=O)C1=C(N=CN1)C(=O)OC

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 184.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl 1H-Imidazole-4,5-dicarboxylate
EINECSNumber (5)-6924
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.