CAS RN 330461-64-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ SIRT2 Inhibitor II, AK-1
The SIRT2 Inhibitor II, AK-1, also referenced under CAS 330461-64-8, controls the biological activity of SIRT2. This small molecule/inhibitor is primarily used for Cell Structure applications.