Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ SIRT2 Inhibitor II, AK-1

Catalog No. 56633110MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-633-110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-633-110MG Supplier MilliporeSigma™ Supplier No. 56633110MG
Add to Cart
Add to Cart

The SIRT2 Inhibitor II, AK-1, also referenced under CAS 330461-64-8, controls the biological activity of SIRT2. This small molecule/inhibitor is primarily used for Cell Structure applications.

A cell permeable, benzylsulfonamide that inhibits SIRT2 activity (IC50= 12.5μM through enzymatic assay) by targeting the SIRT2 nicotinamide binding site, but may also target SIRT1 and SIRT3 (IC50 >40μM) with much less potency. Western analysis illustrates that treatment with AK-1 (> 25μM) leads to an increase in acetylated tubulin. AK-1 (1 to 10μM) is shown to rescue neuronal dysfunction in two in vivo models of Huntington's Disease (HD) in Dropshila and Caenorhabditis elegans. It also demonstrates recovery of neuronal toxicity in a primary model of HD from 1μM to 4μM similar to that of genetic SIRT2 inhibition, and also correlates with the negative regulation of sterol biosynthesis.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description >95% by HPLC
CAS 330461-64-8
Molecular Formula C19H21N3O5S
Formula Weight 403.45g/mol
Solubility DMSO
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.