accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 330581-20-9

IUPAC Name: disodium 4-hydroxynaphthalene-2,7-disulfonate hydrate
Synonyms: disodium 4-hydroxynaphthalene-2,7-disulfonate hydrate
Molecular Weight (g/mol): 366.27
Molecular Formula: C10H8Na2O8S2
InChi Key: BJNIHPYLVMUUPO-UHFFFAOYSA-L
SMILES: O.[Na+].[Na+].OC1=CC(=CC2=CC(=CC=C12)S([O-])(=O)=O)S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)