Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disodium 1-Naphthol-3,6-disulfonate Hydrate 50.0+%, TCI America™
SDP

Catalog No. V003725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V003725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V003725G Supplier TCI America Supplier No. V003725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 330581-20-9
Molecular Formula C10H8Na2O8S2
Molecular Weight (g/mol) 366.266
MDL Number MFCD00003966
InChI Key BJNIHPYLVMUUPO-UHFFFAOYSA-L
Synonym Disodium 4-Hydroxy-2,7-naphthalenedisulfonate, 4-Hydroxy-2,7-naphthalenedisulfonic Acid Disodium Salt, 1-Naphthol-3,6-disulfonic Acid Disodium Salt, Rudolf Guercke Acid Disodium Salt, Violet Acid Disodium Salt
PubChem CID 17749106
IUPAC Name disodium;4-hydroxynaphthalene-2,7-disulfonate;hydrate
SMILES C1=CC2=C(C=C(C=C2C=C1S(=O)(=O)[O-])S(=O)(=O)[O-])O.O.[Na+].[Na+]

Specifications

Color Gray-Red
Quantity 25 g
Formula Weight 348.25
Physical Form Crystalline Powder
Chemical Name or Material Disodium 1-Naphthol-3,6-disulfonate Hydrate
EINECSNumber (4)-0496
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.