accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONNNOOH3COOCH3

CAS RN 330680-46-1

IUPAC Name: 4,4'-dimethyl 6-[4-(methoxycarbonyl)pyridin-2-yl]-[2,2'-bipyridine]-4,4'-dicarboxylate
Synonyms: 4,4'-dimethyl 6-[4-(methoxycarbonyl)pyridin-2-yl]-[2,2'-bipyridine]-4,4'-dicarboxylate
Molecular Weight (g/mol): 407.38
Molecular Formula: C21H17N3O6
InChi Key: YDXTYOYBYASPDS-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=NC=C1)C1=CC(=CC(=N1)C1=NC=CC(=C1)C(=O)OC)C(=O)OC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results