Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl 2,2':6',2″-Terpyridine-4,4',4″-tricarboxylate 96.0+%, TCI America™
SDP

Catalog No. T2959200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2959200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T2959200MG Supplier TCI America Supplier No. T2959200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 330680-46-1
Molecular Formula C21H17N3O6
Molecular Weight (g/mol) 407.382
MDL Number MFCD11042545
InChI Key YDXTYOYBYASPDS-UHFFFAOYSA-N
Synonym 2,2′C:6′C,2′C′C-Terpyridine-4,4′C,4′C′C-tricarboxylic Acid Trimethyl Ester
PubChem CID 66776557
IUPAC Name methyl 2,6-bis(4-methoxycarbonylpyridin-2-yl)pyridine-4-carboxylate
SMILES COC(=O)C1=CC(=NC=C1)C2=CC(=CC(=N2)C3=NC=CC(=C3)C(=O)OC)C(=O)OC

Specifications

Melting Point 241°C
Color White-Yellow
Quantity 200 mg
Formula Weight 407.38
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Trimethyl 2,2′:6′,2′′-Terpyridine-4,4′,4′′-tricarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.