accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3

CAS RN 33083-83-9

IUPAC Name: undecan-5-one
Synonyms: undecan-5-one
Molecular Weight (g/mol): 170.3
Molecular Formula: C11H22O
InChi Key: WENNKWXPAWNIOO-UHFFFAOYSA-N
SMILES: CCCCCCC(=O)CCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results