Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Undecanone 96.0+%, TCI America™
SDP

Catalog No. U004225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U004225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. U004225ML Supplier TCI America Supplier No. U004225ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 33083-83-9
Molecular Formula C11H22O
Molecular Weight (g/mol) 170.30
MDL Number MFCD00048899
InChI Key WENNKWXPAWNIOO-UHFFFAOYSA-N
Synonym 5-undecanone, butyl hexyl ketone, di-n-pentyl ketone, acmc-209hyg, n-butyl n-hexyl ketone
PubChem CID 98677
IUPAC Name undecan-5-one
SMILES CCCCCCC(=O)CCCC

Specifications

Color Yellow
Boiling Point 106°C
Quantity 25 mL
Formula Weight 170.30
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Undecanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.