accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSCH3

CAS RN 3312-77-4

IUPAC Name: 1-(hexadecylsulfanyl)hexadecane
Synonyms: 1-(hexadecylsulfanyl)hexadecane
Molecular Weight (g/mol): 482.94
Molecular Formula: C32H66S
InChi Key: OILQNNHOQFRDJH-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCSCCCCCCCCCCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results