Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexadecyl Sulfide 97.0+%, TCI America™
SDP

Catalog No. H048625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H048625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H048625G Supplier TCI America Supplier No. H048625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3312-77-4
Molecular Formula C32H66S
Molecular Weight (g/mol) 482.94
MDL Number MFCD00026657
InChI Key OILQNNHOQFRDJH-UHFFFAOYSA-N
Synonym Cetyl Sulfide, Dihexadecyl Sulfide
PubChem CID 76823
IUPAC Name 1-hexadecylsulfanylhexadecane
SMILES CCCCCCCCCCCCCCCCSCCCCCCCCCCCCCCCC

Specifications

Melting Point 62°C
Color White
Quantity 25 g
Formula Weight 482.94
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Hexadecyl Sulfide
EINECSNumber (2)-0473&(2)-0475
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.