CAS RN 331465-71-5
IUPAC Name:
2,8,9-tris(2-methylpropyl)-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecan-5-ium
Synonyms:
2,8,9-tris(2-methylpropyl)-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecan-5-ium
Molecular Weight (g/mol):
343.52
Molecular Formula:
C18H40N4P
InChi Key:
WFHPXSHLCFHEIA-UHFFFAOYSA-O
SMILES:
CC(C)CN1CC[NH+]2CCN(CC(C)C)P1N(CC(C)C)CC2
1
–
1
of
1
results
1
Sigma Aldrich 2,8,9-Triisobutyl-2,5,8,9-Tetraaza-1-Phosphabicyclo[3.3.3]Undecane
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.