Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,8,9-Triisobutyl-2,5,8,9-Tetraaza-1-Phosphabicyclo[3.3.3]Undecane

Catalog No. 5658815G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-588-15G 5 g
56-588-11G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 56-588-15G Supplier Sigma Aldrich Supplier No. 5658815G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Together with Pd2(dba)3 (Catalog Number 328774) forms a highly efficient catalyst for one-pot syntheses of trans-4-N,N-diarylaminostilbenes and N,N-diarylaminostyrenes. Catalyst for methanolic cleavage of heteroaromatic carbamates.

Specifications

CAS 331465-71-5
Density 0.964 g/mL at 25°C
Molecular Formula C18H39N4P
Refractive Index n20/D 1.5020
Linear Formula C18H39N4P
Quantity 5 g
MDL Number MFCD03840347
Synonym Triisobutylphosphatrane
Molecular Weight (g/mol) 342.5 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.