CAS RN 33166-77-7
IUPAC Name:
ethyl 3-(3-fluorophenyl)-3-oxopropanoate
Synonyms:
ethyl 3-(3-fluorophenyl)-3-oxopropanoate
Molecular Weight (g/mol):
210.2
Molecular Formula:
C11H11FO3
InChi Key:
MLABEWHVTXMKHP-UHFFFAOYSA-N
SMILES:
CCOC(=O)CC(=O)C1=CC(F)=CC=C1
1
–
1
of
1
results
1
Promotions Available
Ethyl (3-fluorobenzoyl)acetate, 98%
Ethyl (3-fluorobenzoyl)acetate, 98%
CAS: 33166-77-7 Molecular Formula: C11H11FO3 Molecular Weight (g/mol): 210.204 MDL Number: MFCD03424809 InChI Key: MLABEWHVTXMKHP-UHFFFAOYSA-N PubChem CID: 2063904 IUPAC Name: ethyl 3-(3-fluorophenyl)-3-oxopropanoate SMILES: CCOC(=O)CC(=O)C1=CC(=CC=C1)F