Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl (3-fluorobenzoyl)acetate, 98%

Catalog No. AAH6131506
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6131506 5 g
AAH61315MD 250 mg
AAH6131503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6131506 Supplier Thermo Scientific Chemicals Supplier No. H6131506
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 33166-77-7 | C11H11FO3 | 210.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33166-77-7
Molecular Formula C11H11FO3
Molecular Weight (g/mol) 210.204
MDL Number MFCD03424809
InChI Key MLABEWHVTXMKHP-UHFFFAOYSA-N
PubChem CID 2063904
IUPAC Name ethyl 3-(3-fluorophenyl)-3-oxopropanoate
SMILES CCOC(=O)CC(=O)C1=CC(=CC=C1)F

Specifications

Color Colorless to Orange
Density 1.166 g/mL
Boiling Point 262°C to 263°C
Refractive Index 1.517
Quantity 5 g
Synonym ethyl 3-3-fluorophenyl-3-oxopropanoate, ethyl 3-fluorobenzoyl acetate, 3-3-fluoro-phenyl-3-oxo-propionic acid ethyl ester, ethyl 3-3-fluoro-phenyl-3-oxopropanoate, benzenepropanoic acid, 3-fluoro-beta-oxo-, ethyl ester, ethyl3-3-fluorophenyl-3-oxopropanoate, acmc-1cl0f, ethyl-m-fluorobenzoylacetate, ethyl 3'-fluorobenzoylacetate, ksc570a6p
Formula Weight 210.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl (3-fluorobenzoyl)acetate
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.