accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NNH

CAS RN 331717-46-5

IUPAC Name: 1H-perimidin-2-amine
Synonyms: 1H-perimidin-2-amine
Molecular Weight (g/mol): 183.21
Molecular Formula: C11H9N3
InChi Key: UNDUSVBXIVZGOQ-UHFFFAOYSA-N
SMILES: NC1=NC2=CC=CC3=CC=CC(N1)=C23
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)