Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H-Perimidin-2-amine hydrobromide, 97%, Thermo Scientific™

Catalog No. AC213660050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC213660050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC213660050 Supplier Thermo Scientific Chemicals Supplier No. 213660050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 331717-46-5
Molecular Formula C11H12BrN3O
Molecular Weight (g/mol) 282.141
MDL Number MFCD00012749
InChI Key OOXFTZVZWUMSIW-UHFFFAOYSA-N
Synonym 1h-perimidin-2-amine hydrobromide hydrate, 2-aminoperimidine hydrobromide hydrate, 1h-perimidin-2-amine hydrate hydrobromide, 1h-perimidin-2-amine,hydrobromide, hydrate 1:1:1, c11h9n3.hbr.h2o, 2-amino-perinidin-hydrobromide, perimidine-2-ylamine, bromide, hydrate, 2,4-diazatricyclo 7.3.1.0^ 5,13 trideca-1 12 ,2,5,7,9 13 ,10-hexaen-3-amine hydrate hydrobromide
PubChem CID 2795579
IUPAC Name 1H-perimidin-2-amine;hydrate;hydrobromide
SMILES C1=CC2=C3C(=C1)NC(=NC3=CC=C2)N.O.Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 5 g
Formula Weight 282.15
Percent Purity ≥97.5% (on dry substance)
Physical Form Crystalline Powder
Chemical Name or Material 1H-perimidin-2-amine hydrobromide hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.