accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3OOOOH3CH3COOCH3CH3

CAS RN 3319-31-1

IUPAC Name: 1,2,4-tris(2-ethylhexyl) benzene-1,2,4-tricarboxylate
Synonyms: triethylhexyl trimellitate
Molecular Weight (g/mol): 546.79
Molecular Formula: C33H54O6
InChi Key: KRADHMIOFJQKEZ-UHFFFAOYNA-N
SMILES: CCCCC(CC)COC(=O)C1=CC=C(C(=O)OCC(CC)CCCC)C(=C1)C(=O)OCC(CC)CCCC
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results