Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trioctyl trimellitate

Catalog No. 5381404L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-814-04L 4 L
53-814-01L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 53-814-04L Supplier Sigma Aldrich Supplier No. 5381404L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trioctyl trimellitate (TOTM) is a flammable plasticizer used to improve the flexibility and durability of poly(vinyl chloride) (PVC).TOTM can plasticize PVC to form a nanocomposite with multi-walled carbon nanotube which can be potentially used for anti-static applications.

Specifications

CAS 3319-31-1
Density 0.99 g/mL (at 25°C)
Boiling Point 414°C
Molecular Formula C33H54O6
Refractive Index n20/D 1.485 (literature)
Linear Formula C6H3-1,2,4-[CO2CH2CH(C2H5)(CH2)3 CH3]3
Quantity 4 L
MDL Number MFCD00053799
Synonym TOTM; Tris(2-ethylhexyl) trimellitate
Molecular Weight (g/mol) 546.78
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.