accessibility menu, dialog, popup

UserName

Viewing profile as user:
FCl

CAS RN 332-43-4

IUPAC Name: 1-(2-chloroethyl)-4-fluorobenzene
Synonyms: 1-(2-chloroethyl)-4-fluorobenzene
Molecular Weight (g/mol): 158.6
Molecular Formula: C8H8ClF
InChi Key: LPUURRVXQCVXCX-UHFFFAOYSA-N
SMILES: FC1=CC=C(CCCl)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results