accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 332154-57-1

IUPAC Name: [4-(carbonochloridoyl)phenyl]boronic acid
Synonyms: 4-(carbonochloridoyl)phenylboronic acid
Molecular Weight (g/mol): 184.38
Molecular Formula: C7H6BClO3
InChi Key: ARKQVAQLGLIPAU-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results