Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorocarbonylbenzeneboronic anhydride, tech. 90%, Thermo Scientific™

Catalog No. AAH5285203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5285203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5285203 Supplier Thermo Scientific Chemicals Supplier No. H5285203

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 332154-57-1
Molecular Formula C7H6BClO3
Molecular Weight (g/mol) 184.382
MDL Number MFCD06801674
InChI Key ARKQVAQLGLIPAU-UHFFFAOYSA-N
Synonym 4-chlorocarbonylphenylboronic acid, benzoylchloride-4-boronic acid, 4-chlorocarbonyl phenyl boronic acid, boronic acid, b-4-chlorocarbonyl phenyl, 4-chlorocarbonylphenyl boronic anhydride, p-chlorocarbonylphenylboronic acid, 4-chlorocarbonylphenylboronic boronic acid, 4-carbonochloridoyl phenylboronic acid, pubchem9545
PubChem CID 15345913
IUPAC Name (4-carbonochloridoylphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)C(=O)Cl)(O)O

Specifications

Melting Point >300°C
Quantity 1 g
Formula Weight 166.37
Percent Purity 90%
Grade Technical
Chemical Name or Material 4-Chlorocarbonylbenzeneboronic anhydride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.