accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33229-34-4

IUPAC Name: 2-({4-[bis(2-hydroxyethyl)amino]-2-nitrophenyl}amino)ethan-1-ol
Synonyms: HC blue NO. 2
Molecular Weight (g/mol): 285.3
Molecular Formula: C12H19N3O5
InChi Key: MIWUTEVJIISHCP-UHFFFAOYSA-N
SMILES: OCCNC1=CC=C(C=C1[N+]([O-])=O)N(CCO)CCO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results