Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-[4-(2-Hydroxyethylamino)-3-nitrophenylimino]diethanol 98.0+%, TCI America™
SDP

Catalog No. T27875G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27875G 5 g
T278725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27875G Supplier TCI America Supplier No. T27875G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 33229-34-4
Molecular Formula C12H19N3O5
Molecular Weight (g/mol) 285.30
MDL Number MFCD00071762
InChI Key MIWUTEVJIISHCP-UHFFFAOYSA-N
Synonym N1,N4,N4-Tris(2-hydroxyethyl)-2-nitro-1,4-phenylenediamine
PubChem CID 36383
ChEBI CHEBI:82472
IUPAC Name 2-({4-[bis(2-hydroxyethyl)amino]-2-nitrophenyl}amino)ethan-1-ol
SMILES OCCNC1=CC=C(C=C1[N+]([O-])=O)N(CCO)CCO

Specifications

Melting Point 108°C
Quantity 5 g
Formula Weight 285.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-[4-(2-Hydroxyethylamino)-3-nitrophenylimino]diethanol
RTECSNumber KL2880000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.