accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNSOHO

CAS RN 332345-29-6

IUPAC Name: 2-(morpholin-4-yl)-1,3-thiazole-5-carboxylic acid
Synonyms: 2-(morpholin-4-yl)-1,3-thiazole-5-carboxylic acid
Molecular Weight (g/mol): 214.24
Molecular Formula: C8H10N2O3S
InChi Key: NILKOMDINYFEEX-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C(S1)N1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results