Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Morpholino-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC62401DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC62401DE 5 g Amber Glass Bottle
CC62401CB 250 mg Amber Glass Bottle
CC62401DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC62401DE Supplier Thermo Scientific Chemicals Supplier No. CC62401DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 332345-29-6
Molecular Formula C8H10N2O3S
Molecular Weight (g/mol) 214.239
InChI Key NILKOMDINYFEEX-UHFFFAOYSA-N
Synonym 2-morpholino-1,3-thiazole-5-carboxylic acid, 2-morpholin-4-yl-1,3-thiazole-5-carboxylic acid, 2-morpholin-4-yl-thiazole-5-carboxylic acid, 5-thiazolecarboxylic acid, 2-4-morpholinyl
PubChem CID 15475835
IUPAC Name 2-morpholin-4-yl-1,3-thiazole-5-carboxylic acid
SMILES C1COCCN1C2=NC=C(S2)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 214.24
Percent Purity 97%
Chemical Name or Material 2-morpholino-1, 3-thiazole-5-carboxylic acid
Hazard Category GHS Signal Word: Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.