accessibility menu, dialog, popup

UserName

Viewing profile as user:
SSOOOOOOOHOHNaNa

CAS RN 332360-05-1

IUPAC Name: disodium 1,2-dihydroxyethane-1,2-disulfonate
Synonyms: disodium 1,2-dihydroxyethane-1,2-disulfonate
Molecular Weight (g/mol): 266.15
Molecular Formula: C2H4Na2O8S2
InChi Key: BXUKAXFDABMVND-UHFFFAOYNA-L
SMILES: [Na+].[Na+].OC(C(O)S([O-])(=O)=O)S([O-])(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results