accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNClNOOFFFNOO

CAS RN 33245-39-5

IUPAC Name: N-(2-chloroethyl)-2,6-dinitro-N-propyl-4-(trifluoromethyl)aniline
Synonyms: fluchloralin
Molecular Weight (g/mol): 355.7
Molecular Formula: C12H13ClF3N3O4
InChi Key: MNFMIVVPXOGUMX-UHFFFAOYSA-N
SMILES: CCCN(CCCl)C1=C(C=C(C=C1[N+]([O-])=O)C(F)(F)F)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results