accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33252-29-8

IUPAC Name: 6-chloropyridine-2-carbonitrile
Synonyms: 6-chloropyridine-2-carbonitrile
Molecular Weight (g/mol): 138.55
Molecular Formula: C6H3ClN2
InChi Key: PGZHSVWXFKKCNR-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(=N1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results